missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium dihydrogen phosphate, Puratronic™, 99.999% (metals basis), Si 10ppm max

Manufacturer:  Alfa Aesar™ 1084014

 View more versions of this product

Catalog No. AA1084014



Specifications

Specifications

Potassium dihydrogen phosphate
(metals basis)
KH2PO4
14,7659
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium;dihydrogen phosphate
516951
136.09
99.999%
Si 10ppm max.
Hygroscopic
253°C
7778-77-0
H2KO4P
MFCD00011401
monobasic potassium phosphate
OP(=O)(O)[O-].[K+]
136.084
CHEBI:63036
Crystalline Powder
Puratronic™
2.338g/mL
White
25g

Chemical Identifiers

7778-77-0
136.084
GNSKLFRGEWLPPA-UHFFFAOYSA-M
516951
potassium;dihydrogen phosphate
H2KO4P
MFCD00011401
monobasic potassium phosphate
CHEBI:63036
OP(=O)(O)[O-].[K+]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : TC6615500

TSCA : TSCA

Documents