missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium dihydrogen phosphate, ACS, 99.0% min., Alfa Aesar™

For Use in Buffers For pH Determination

Manufacturer:  Alfa Aesar 1159436

 View more versions of this product

Catalog No. AA1159436



Specifications

Specifications

Potassium dihydrogen phosphate
7778-77-0
H2KO4P
White crystals
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium;dihydrogen phosphate
516951
136.09
ACS
500g
For analysis ACS
99.0% min.
MFCD00011401
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
OP(=O)(O)[O-].[K+]
136.084
CHEBI:63036
Crystalline
White

Chemical Identifiers

7778-77-0
136.084
GNSKLFRGEWLPPA-UHFFFAOYSA-M
516951
potassium;dihydrogen phosphate
H2KO4P
MFCD00011401
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
CHEBI:63036
OP(=O)(O)[O-].[K+]
Safety and Handling

Safety and Handling

H315-H319: Causes skin irritation. Causes serious eye irritation.

EINECSNumber : 231-913-4

Documents