missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium dihydrogen citrate hydrate, 99% (dry basis)

Manufacturer:  Alfa Aesar™ 44415A3

 View more versions of this product

Catalog No. AA44415A3



Specifications

Specifications

Potassium dihydrogen citrate hydrate
(dry basis)
MFCD00036472
14,7624
Soluble in water. Slightly soluble in ethanol
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.[K]
231.221
230.22 (Anhydrous)
99%
2kg
866-83-1
C6H8KO7
4772973
potassium dihydrogen citrate
NAVWVHRQSDHCHD-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid;potassium
73896894
Powder/ Chunks
Hygroscopic

Chemical Identifiers

866-83-1
231.221
NAVWVHRQSDHCHD-UHFFFAOYSA-N
73896894
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.[K]
C6H8KO7
MFCD00036472
potassium dihydrogen citrate
2-hydroxypropane-1,2,3-tricarboxylic acid;potassium
Safety and Handling

Safety and Handling

EINECSNumber : 212-753-4

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents