missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium dichromate, MilliporeSigma™

For analysis EMSURE™ ACS, ISO, Reag. Ph Eur

Manufacturer:  MilliporeSigma 1048640500

 View more versions of this product

Catalog No. M1048640500



Specifications

Specifications

Potassium dichromate
Cr2K2O7
Soluble in water: 130g/L at 10°C
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
294.182
CHEBI:53444
2.7g/cm3 at 20°C
Plastic Bottle
>500°C
7778-50-9
bichromate of potash
KMUONIBRACKNSN-UHFFFAOYSA-N
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
24502
294.19
Odorless
ACS, ISO, Reag. Ph Eur
500g

Chemical Identifiers

7778-50-9
294.182
bichromate of potash
CHEBI:53444
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
Cr2K2O7
KMUONIBRACKNSN-UHFFFAOYSA-N
24502
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
Safety and Handling

Safety and Handling

H350: May cause cancer.

H360FD: May damage fertility. May damage the unborn child.

H340: May cause genetic defects.

EINECSNumber : 231-906-6

RTECSNumber : HX7680000

Recommended Storage : Store at 5° to 30°C (41° to 86°F), Tightly closed. Dry. Do not store near combustible material. Keep locked up or in an area accessible only to qualified or authorized persons.

SDS Documents