missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Dichromate (Crystalline/Certified ACS), Fisher Chemical

Manufacturer:  Thermo Scientific P1883

 View more versions of this product

Catalog No. P188-3



Specifications

Specifications

Potassium Dichromate
Certified ACS Grade
≥99%
MFCD00011367
Potassium Bichromate
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
294.182
CHEBI:53444
Solid
0.001% max.
Pass Test
0.001% max.
Poly Bottle
0.005% max.
500°C
398°C
3kg
7778-50-9
≥99%
Cr2K2O7
3086
KMUONIBRACKNSN-UHFFFAOYSA-N
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
24502
294.18
0.003% max.
2.676g/cm³
0.005% max.
0.05% max.
0.02% max.
>500°C
Orange
4

Chemical Identifiers

7778-50-9
294.182
KMUONIBRACKNSN-UHFFFAOYSA-N
24502
Cr2K2O7
MFCD00011367
Potassium Bichromate
CHEBI:53444
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 6.1, : Poison

SDS Documents