Please login to your online account to display your discounted pricing

Potassium Dichromate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Quantity 125g
Packaging Poly Bottle

Manufacturer: avantor performance  309004

 View all options for this product

Catalog No. 02003753

 

Description & Specifications

Chemical Identifiers

Packaging Poly Bottle
Quantity 125g
Specific Gravity 1L = 2.68kg
Quantity 125g
Chemical Name or Material Potassium Dichromate
CAS 7778-50-9
Purity Grade BAKER ANALYZED™ ACS Reagent
Physical Form Crystal
Molecular Formula K2Cr2O7
Formula Weight 294.19g/mol

Specifications

Chemical Name or Material Potassium Dichromate
CAS 7778-50-9
Purity Grade BAKER ANALYZED™ ACS Reagent
Molecular Formula K2Cr2O7
Formula Weight 294.19g/mol