Please login to your online account to display your discounted pricing

Potassium Dichromate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$347.37 - $1,600.56

Chemical Identifiers

Chemical Name or Material Potassium Dichromate
CAS 7778-50-9
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula K2Cr2O7
Formula Weight 294.19
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003754

 
avantor performance
309005
Poly Bottle 2.5kg Each for $1,600.56

02003755

 
avantor performance
309001
Poly Bottle 500g Each for $347.37
Description & Specifications

Specifications

Formula Weight 294.19
Physical Form Crystals
Purity Grade Baker Analyzed™ ACS Reagent
Chemical Name or Material Potassium Dichromate
CAS 7778-50-9
Molecular Formula K2Cr2O7
Quantity 2.5kg
Packaging Poly Bottle
Specific Gravity 1L = 2.68kg