Please login to your online account to display your discounted pricing

Potassium Dichromate, Certified, 9.0% (w/v) ±0.2% (w/v), LabChem™

$48.69 - $48.69

Chemical Identifiers

Chemical Name or Material Potassium Dichromate
CAS 7732-18-5
CAS Max % 91.5
CAS 7778-50-9
CAS Max % 8.5
Purity Grade Certified Grade
Linear Formula K2Cr2O7
Formula Weight 294.18
UN Number UN3122
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC189471

 
lab chem
LC189471
500mL Each for $48.69
Description & Specifications

Specifications

Formula Weight 294.18
Purity Grade Certified Grade
Assay 9.0% ± 0.2% (w/v)
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist, vapors, spray.
Do not get in eyes, on skin, or on clothing.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Decomposition Information Contains hexavalent Chromium
Linear Formula K2Cr2O7
Chemical Name or Material Potassium Dichromate
Viscosity 0.954cSt
CAS 7732-18-5
Physical Form Liquid
CAS Max % 91.5
CAS 7778-50-9
CAS Max % 8.5
Identification Passes Test
Specific Gravity 1.059g/mL
Storage Information Room Temperature
Density 1.059g/mL
Solubility Soluble in water
UN Number UN3122
Packaging Poly Bottle
Health Hazard 1 Danger
Color Clear, orange
Quantity 500mL
Health Hazard 2 GHS H Statement
Toxic if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs (liver, kidneys, skin) through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist, vapors, spray.
Do not get in eyes, on skin, or on clothing.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Toxic if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs (liver, kidneys, skin) through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.