Please login to your online account to display your discounted pricing

Potassium Dichromate ACS, Ricca Chemical


Quantity 500g
Packaging Poly wht WM

Manufacturer: ricca chemical company  RIRDCP0420500B1

 View more versions of this product

Catalog No. RDCP0420500

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500g
Purity Grade ACS Grade
Physical Form Powder
Color Orange-red
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material Potassium Dichromate
Shelf Life 60 months
CAS 7778-50-9
Packaging Poly wht WM

Specifications

Chemical Name or Material Potassium Dichromate
CAS 7778-50-9
CAS Min % 98.0
CAS Max % 100.0
Purity Grade ACS Grade
Physical Form Powder
Accessories

60 months