Please login to your online account to display your discounted pricing

Potassium Dichromate, ACS Grade, ≥ 99.0%, LabChem™

$172.80 - $172.80

Chemical Identifiers

Chemical Name or Material Potassium Dichromate
CAS 7778-50-9
CAS Max % 100
Purity Grade ACS Grade
Linear Formula K2Cr2O7
Formula Weight 294.18
UN Number UN3086
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC189401

 
lab chem
LC189401
500g Each for $172.80
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 294.18
Boiling Point 500°C
Quantity 500g
Identification Passes Test
Storage Information Room Temperature
Packaging Poly Bottle
Color Orange
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Do not breathe dust.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up in a well ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Purity Grade ACS Grade
UN Number UN3086
Melting Point 398°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
May intensify fire; oxidizer.
Fatal if inhaled.
Toxic if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
CAS 7778-50-9
Physical Form Powder
CAS Max % 100
Solubility Soluble in water
Linear Formula K2Cr2O7
Chemical Name or Material Potassium Dichromate
Decomposition Information Contains hexavalent Chromium
EINECS Number 231-906-6
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Do not breathe dust.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up in a well ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May intensify fire; oxidizer.
Fatal if inhaled.
Toxic if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.