Learn More
Potassium dichromate, 99.5%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 196595000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
500.0°C | |
398.0°C | |
7778-50-9 | |
Fine Crystals or Crystalline Powder | |
99.5% min. (Iodometry) | |
K2Cr2O7 | |
08,410; 09,383; 10,324; 12,405 | |
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash | |
KMUONIBRACKNSN-UHFFFAOYSA-N | |
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium | |
24502 | |
294.19 | |
Analytical | |
Plastic bottle |
Potassium dichromate | |
p.a. | |
Orange | |
500 g | |
Cr2K2O7 | |
MFCD00011367 | |
15, 7748 | |
Solubility in water: 125g/L (20°C). Other solubilities: 50 g/100 mL water (100°C), insoluble in alcohol | |
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] | |
294.19 | |
CHEBI:53444 | |
99.5% | |
0.05% max. (105°C) |
Chemical Identifiers
7778-50-9 | |
294.19 | |
KMUONIBRACKNSN-UHFFFAOYSA-N | |
24502 | |
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
Cr2K2O7 | |
MFCD00011367 | |
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash | |
CHEBI:53444 | |
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficu
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major
GHS Signal Word: Danger
EINECSNumber : 231-906-6
RTECSNumber : HX7680000
TSCA : TSCA
RUO – Research Use Only