Please login to your online account to display your discounted pricing

Potassium dichromate, 99.5%, for analysis, ACROS Organics™


Quantity 50g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  196590500

 View more versions of this product

Catalog No. AC196590500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.5%
Physical Form Fine Crystalline Powder or Crystals
Formula Weight 294.19
Quantity 50g
Color Orange
EINECS Number 231-906-6
TSCA TSCA
Purity Grade For analysis
RTECS Number HX7680000
Fieser 08,410; 09,383; 10,324; 12,405
Merck Index 15, 7748
Loss on Drying 0.05% max. (105°C)
Sodium (Na) 200ppm max.
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement
May cause an allergic skin reaction.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
Harmful in contact with skin.
Causes damage to organs through prolonged or repeated exposure.
May damage fertility. May damage the unborn child.
Toxic if swallowed.
May intensify fire; oxidizer.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
Take any precaution to avoid mixing with combustibles.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Potassium dichromate
Name Note p.a.
Assay Percent Range 99.5%
Molecular Formula Cr2K2O7
Linear Formula K2Cr2O7
MDL Number MFCD00011367
CAS 7778-50-9
Iron (Fe) 20ppm max.
Lead (Pb) 50ppm max.
Packaging Plastic bottle
Sulfate 50ppm max.
Calcium (Ca) 20ppm max.
Chlorine (Cl) 10ppm max.
Copper (Cu) 10ppm max.

Specifications

Chemical Name or Material Potassium dichromate
Name Note p.a.
CAS 7778-50-9
Purity Grade For analysis
Assay ≥99.5%
Assay Percent Range 99.5%
Molecular Formula Cr2K2O7
Linear Formula K2Cr2O7
Formula Weight 294.19
MDL Number MFCD00011367
Accessories

Danger

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
Take any precaution to avoid mixing with combustibles.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
May cause an allergic skin reaction.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
Harmful in contact with skin.
Causes damage to organs through prolonged or repeated exposure.
May damage fertility. May damage the unborn child.
Toxic if swallowed.
May intensify fire; oxidizer.
May cause respiratory irritation.

SDS & Safety Documents