missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Potassium Dichromate, 0.500 N (N/2), Ricca Chemical
Chemical Identifiers
CAS |
7778-50-9 |
Molecular Formula |
Cr2K2O7 |
Molecular Weight (g/mol) |
294.182 |
InChI Key |
KMUONIBRACKNSN-UHFFFAOYSA-N |
Synonym |
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash |
PubChem CID |
24502 |
ChEBI |
CHEBI:53444 |
IUPAC Name |
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
SMILES |
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] |
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
|
7778-50-9 |
|
294.182 |
|
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash |
|
CHEBI:53444 |
|
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] |
|
Cr2K2O7 |
|
KMUONIBRACKNSN-UHFFFAOYSA-N |
|
24502 |
|
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
Specifications
|
Orange |
|
96.0,2.35 |
|
Cr2K2O7 |
|
KMUONIBRACKNSN-UHFFFAOYSA-N |
|
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
|
24502 |
|
Natural Poly Bottle |
|
Liquid |
|
Potassium Dichromate |
|
7778-50-9,7732-18-5 |
|
100.0,2.44 |
|
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash |
|
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] |
|
294.182 |
|
CHEBI:53444 |
|
500 mL |
|
0.500Normal |
SDS
missing translation for 'documents'