Please login to your online account to display your discounted pricing

Potassium Dichromate, 0.250 Normal, 0.04167 Molar, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Potassium Dichromate
CAS 7732-18-5
CAS Min % 97.0
CAS Max % 100.0
CAS 7778-50-9
CAS Min % 1.19
CAS Max % 1.24
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

60702-5

 
ricca chemical company
RI60702.5
Cubitainer 10L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

60705

  SDS
ricca chemical company
RI60705
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear orange
Quantity 10L
CAS 7732-18-5
CAS Max % 1.24
Concentration or Composition Notes 0.250Normal,0.04167Molar
CAS Min % 97.0
CAS Max % 100.0
CAS 7778-50-9
CAS Min % 1.19
Packaging Cubitainer
Chemical Name or Material Potassium Dichromate