Please login to your online account to display your discounted pricing

Potassium Dichromate, 0.100 Normal, 0.0167 Molar, Ricca Chemical


Quantity 100mL
Packaging Poly natural

Manufacturer: ricca chemical company  RIR6060000100A

 View more versions of this product

Catalog No. R6060000100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Color Clear orange
Physical Form Liquid
Quantity 100mL
Chemical Name or Material Potassium Dichromate
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 7778-50-9
Packaging Poly natural
CAS Min % 0.48
CAS Max % 0.5
Methods ACS/EP/USP Volumetric Solution
Concentration or Composition Notes 0.100N,0.0167M

Specifications

Chemical Name or Material Potassium Dichromate
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 7778-50-9
CAS Min % 0.48
CAS Max % 0.5
Physical Form Liquid