missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium cyclopropyltrifluoroborate, tech. 90%

Chemical Identifiers

CAS 1065010-87-8
Formula Weight 147.98
MDL Number MFCD09265031
Synonym Cyclopropyltrifluoroboric acid potassium salt
Chemical Name or Material Potassium cyclopropyltrifluoroborate
Grade Technical Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6067303 SDS Alfa Aesar™
H6067303
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6067306 SDS Alfa Aesar™
H6067306
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1065010-87-8
MFCD09265031
Potassium cyclopropyltrifluoroborate
147.98
Cyclopropyltrifluoroboric acid potassium salt
Technical Grade

Specifications

348° to 350°C
1065010-87-8
White
MFCD09265031
Powder
90%
147.98
Air Sensitive
Cyclopropyltrifluoroboric acid potassium salt
1g
Potassium cyclopropyltrifluoroborate
Technical Grade

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning