Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium cyclohexanebutyrate, K 16.6-18.8%

$967.34 - $967.34

Chemical Identifiers

Chemical Name or Material Potassium cyclohexanebutyrate
CAS 62638-03-3
Molecular Formula C10H17KO2
Formula Weight 208.33
MDL Number MFCD00013051
Synonym Cyclohexanebutyric acid potassium salt; Potassium 4-cyclohexylbutyrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1783914

  SDS
alfa aesar™
A1783914
25g Each for $967.34
Description & Specifications

Specifications

Formula Weight 208.33
CAS 62638-03-3
Chemical Name or Material Potassium cyclohexanebutyrate
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay Percent Notes K 16.6 to 18.8%
Molecular Formula C10H17KO2
Synonym Cyclohexanebutyric acid potassium salt; Potassium 4-cyclohexylbutyrate
TSCA TSCA
Quantity 25g
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00013051
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.