missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium cyclohexanebutyrate, AAS, K 18.8%

Manufacturer:  Alfa Aesar™ 8726503

 View more versions of this product

Catalog No. AA8726503



Specifications

Specifications

Potassium cyclohexanebutyrate
AAS Grade
MFCD00013051
LBYNWSSEILNGLN-UHFFFAOYSA-M
potassium;4-cyclohexylbutanoate
23692296
Powder
K 18.8%
Hygroscopic
62638-03-3
C10H17KO2
Cyclohexanebutyric acid potassium salt; Potassium 4-cyclohexylbutyrate
C1CCC(CC1)CCCC(=O)[O-].[K+]
208.342
208.33
White
1g

Chemical Identifiers

62638-03-3
208.342
LBYNWSSEILNGLN-UHFFFAOYSA-M
23692296
C1CCC(CC1)CCCC(=O)[O-].[K+]
C10H17KO2
MFCD00013051
Cyclohexanebutyric acid potassium salt; Potassium 4-cyclohexylbutyrate
potassium;4-cyclohexylbutanoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents