missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Cyanide (Granular/Certified), Fisher Chemical

Manufacturer:  Thermo Scientific FLP223I500

 View more versions of this product

Catalog No. P223I-500



Specifications

Specifications

Potassium Cyanide
CKN
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, potassium cyanide k cn, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
[C-]#N.[K+]
65.116
65.12
≥98%
0.5% max.
0.03% max.
Glass Bottle
1.0% max.
0.002% max.
1625°C
634°C
500g
151-50-8
MFCD00011397
NNFCIKHAZHQZJG-UHFFFAOYSA-N
potassium;cyanide
9032
Powder/Solid
Certified
Pass Test
2ppm max.
0.02% max.
0.04% max.
P.T. (about 0.02%)
White
11 to 12

Chemical Identifiers

151-50-8
65.116
NNFCIKHAZHQZJG-UHFFFAOYSA-N
9032
[C-]#N.[K+]
CKN
MFCD00011397
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, potassium cyanide k cn, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
potassium;cyanide
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 6.1, : Poison

SDS Documents