Please login to your online account to display your discounted pricing

Potassium Cyanide (Granular/Certified), Fisher Chemical

$329.69 - $532.08

Chemical Identifiers

Chemical Name or Material Potassium Cyanide
CAS 151-50-8
Purity Grade Certified Grade
Physical Form Powder/Solid
Assay ≥98%
Molecular Formula CKN
Formula Weight 65.12
MDL Number MFCD00011397
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

P223I-100

  SDS
fisher scientific company
FLP223I100
Glass Bottle 100g Each for $329.69

P223I-500

  SDS
fisher scientific company
FLP223I500
Glass Bottle 500g Each for $532.08
Description & Specifications

Specifications

Assay ≥98%
Purity Grade Certified Grade
Physical Form Powder/Solid
Formula Weight 65.12
Boiling Point 1625°C
Melting Point 634°C
Quantity 100g
DOT Information DOT Class 6.1, : Poison
Molecular Formula CKN
Viscosity [NULL]
Color White
pH 11 to 12
Lead (Pb) 2ppm max.
Sodium (Na) 1.0% max.
Chemical Name or Material Potassium Cyanide
Identification Pass Test
Packaging Glass Bottle
Thiocyanate P.T. (about 0.02%)
MDL Number MFCD00011397
CAS 151-50-8
ChemAlert Storage Symbol Blue
Chloride 0.5% max.
Iron (Fe) 0.03% max.
Phosphate 0.02% max.
Sulfate 0.04% max.
Sulfide 0.002% max.
Safety and Handling