Learn More
Potassium cyanide, 97+%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 388315000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
1625.0°C | |
634.0°C | |
Colorless | |
500 g | |
CKN | |
MFCD00011397 | |
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french | |
NNFCIKHAZHQZJG-UHFFFAOYSA-N | |
potassium;cyanide | |
9032 | |
≥97% | |
Plastic bottle |
Potassium cyanide | |
151-50-8 | |
Liquid | |
97% min. (Argentometry) | |
CKN | |
15,7747 | |
Solubility in water: 400g/L (20°C). Other solubilities: ethanol 4.5g/L (19.5°C) | |
[C-]#N.[K+] | |
65.12 | |
65.12 | |
Extra Pure |
Chemical Identifiers
151-50-8 | |
65.12 | |
NNFCIKHAZHQZJG-UHFFFAOYSA-N | |
9032 |
CKN | |
MFCD00011397 | |
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure if inhaled.
Very toxic to aquatic life with
GHS P Statement
Absorb spillage to prevent material damage.
Rinse mouth.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at r
GHS Signal Word: Danger
EINECSNumber : 205-792-3
RUO – Research Use Only