missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium cyanate, 97%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 201115000

 View more versions of this product

Catalog No. AC201115000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium cyanate
Pure
White
500 g
CKNO
MFCD00011411
15, 7746
Solubility in water: 750g/L (20°C). Other solubilities: insoluble in alcohol
[K+].[O-]C#N
81.12
CHEBI:38904
97%
Plastic bottle
315.0°C
590-28-3
Crystalline Powder and/or Chunks
97%
KOCN
01,1059
potassium cyanate, kaliumcyanat, kocn, aero cyanate, cyanic acid, potassium salt, alicyanate, bulpur, potassiumcyanate, weedanol cyanol, caswell no. 688
GKKCIDNWFBPDBW-UHFFFAOYSA-M
potassium;cyanate
11378442
81.12
Pure

Chemical Identifiers

590-28-3
81.12
GKKCIDNWFBPDBW-UHFFFAOYSA-M
11378442
potassium;cyanate
CKNO
MFCD00011411
potassium cyanate, kaliumcyanat, kocn, aero cyanate, cyanic acid, potassium salt, alicyanate, bulpur, potassiumcyanate, weedanol cyanol, caswell no. 688
CHEBI:38904
[K+].[O-]C#N
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-676-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only