missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium citrate tribasic monohydrate, 99%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 244210010

 View more versions of this product

Catalog No. AC244210010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium citrate tribasic monohydrate
180.0°C
Plastic bottle
KO2CCH2C(OH)(CO2K)CH2CO2K·H2O
15,7744
Solubility in water: 640g/L (25°C).
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
324.42
CHEBI:64746
99%
6100-05-6
99%
C6H5K3O7·H2O
1 kg
potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
PJAHUDTUZRZBKM-UHFFFAOYSA-K
tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
2735208
324.42
Extra Pure

Chemical Identifiers

6100-05-6
324.42
potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
CHEBI:64746
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
C6H5K3O7·H2O
PJAHUDTUZRZBKM-UHFFFAOYSA-K
2735208
tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only