missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium citrate tribasic monohydrate, 99%, extra pure, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC244210050
Specifications
Potassium citrate tribasic monohydrate | |
180.0°C | |
6100-05-6 | |
C6H5K3O7·H2O | |
15,7744 | |
Solubility in water: 640g/L (25°C). | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+] | |
324.42 | |
CHEBI:64746 | |
99% |
Plastic bottle | |
5kg | |
99% | |
KO2CCH2C(OH)(CO2K)CH2CO2K·H2O | |
ccris 6543, citric acid tripotassium salt, citric acid, tripotassium salt, monohydrate, ee90oni6ff, potassium citrate monohydrate, potassium citrate tribasic monohydrate, tri-potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, urocit-k | |
PJAHUDTUZRZBKM-UHFFFAOYSA-K | |
tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate | |
2735208 | |
324.42 | |
Extra Pure |
Chemical Identifiers
6100-05-6 | |
324.42 | |
ccris 6543, citric acid tripotassium salt, citric acid, tripotassium salt, monohydrate, ee90oni6ff, potassium citrate monohydrate, potassium citrate tribasic monohydrate, tri-potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, urocit-k | |
CHEBI:64746 | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+] |
C6H5K3O7·H2O | |
PJAHUDTUZRZBKM-UHFFFAOYSA-K | |
2735208 | |
tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate |