missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Citrate Monohydrate (Granular/USP/FCC), Fisher Chemical

Manufacturer:  Thermo Scientific FLP218212

 View more versions of this product

Catalog No. P218-212



Specifications

Specifications

Pass Test (FCC/USP)
Pass Test (FCC/USP)
Poly Bottle
White
2.5kg
6100-05-6
KO2CCH2C(OH)(CO2K)CH2CO2K·H2O
potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
324.41
CHEBI:64746
Solid
FCC/USP
Potassium Citrate Monohydrate
3.0 to 6.0% (FCC/USP)
Pass Test (USP)
180°C
7.5 to 9.0
C6H7K3O8
MFCD00150442
PJAHUDTUZRZBKM-UHFFFAOYSA-K
tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
2735208
324.41
99.0 to 100.5% (USP)

Chemical Identifiers

6100-05-6
324.41
PJAHUDTUZRZBKM-UHFFFAOYSA-K
2735208
tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
C6H7K3O8
MFCD00150442
potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
CHEBI:64746
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
Safety and Handling

Safety and Handling

SDS Documents