missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Citrate, Monohydrate, Granular, U.S.P. - F.C.C., J.T.Baker™

Manufacturer:  Avantor Performance 306801

 View more versions of this product

Catalog No. 02003750



Specifications

Specifications

Potassium Citrate
6100-05-6
potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
324.41
CHEBI:64746
Granular
Wide Mouth Amber Glass Bottle
500g
Monohydrate
C6H7K3O8
PJAHUDTUZRZBKM-UHFFFAOYSA-K
tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
2735208
324.42
FCC/USP
1L = 1.98kg

Chemical Identifiers

6100-05-6
324.41
potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
CHEBI:64746
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
C6H7K3O8
PJAHUDTUZRZBKM-UHFFFAOYSA-K
2735208
tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
Documents