Learn More
Potassium Chromate Indicator, For Chloride, Certified, 5% (w/v), LabChem™
Supplier: LabChem LC188501
Specifications
Potassium Chromate Indicator | |
7732-18-5,7789-00-6,7761-88-8 | |
K2CrO4 | |
Passes Test | |
5% (w/v) | |
Poly Bottle | |
Passes Test | |
1.05g/mL | |
500 mL |
For Chloride | |
94.9992,5,0.0008 | |
Soluble in water | |
194.2 | |
Certified | |
1.05g/mL | |
Contains hexavalent Chromium | |
Orange/Yellow | |
Liquid |
Chemical Identifiers
7732-18-5 |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
May cause genetic defects.
May cause cancer.
May cause damage to organs (blood, kidneys, liver) through prolonged or repeated exposure.
Very toxic to aquatic life.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist.
Wear protective gloves, protective clothing, eye protection, face protection.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Take off contaminated clothing and wash it before reuse.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature