Learn More
Potassium Chromate (Granular/Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific P220500
Specifications
Potassium Chromate | |
7789-00-6 | |
8.6 to 9.8 | |
500 g | |
CrK2O4 | |
3087 | |
XMXNVYPJWBTAHN-UHFFFAOYSA-N | |
dipotassium dioxochromiumbis(olate) | |
24597 | |
194.19 | |
Certified ACS | |
Poly Bottle |
975°C | |
Yellow | |
Solid | |
99.5 to 100.5 % | |
MFCD00011368 | |
potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687 | |
[K+].[K+].[O-][Cr]([O-])(=O)=O | |
194.19 | |
CHEBI:75249 | |
99.5 to 100.5% | |
Pass Test |
Chemical Identifiers
7789-00-6 | |
194.19 | |
XMXNVYPJWBTAHN-UHFFFAOYSA-N | |
24597 | |
dipotassium dioxochromiumbis(olate) |
CrK2O4 | |
MFCD00011368 | |
potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687 | |
CHEBI:75249 | |
[K+].[K+].[O-][Cr]([O-])(=O)=O |
Safety and Handling
DANGER!
Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. May cause cancer by inhalation. May cause heritable genetic damage. Irritating to eyes, respiratory system and skin. May cause an allergic skin reaction. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.
NFPA
Health:2
Flammability:0
Instability:2
DOTInformation : DOT Class 5.1, : Oxidizer