Please login to your online account to display your discounted pricing

Potassium Chromate, For Silica, 0.063%, LabChem™

$46.37 - $46.37

Chemical Identifiers

Chemical Name or Material Potassium Chromate
CAS 7732-18-5
CAS Max % 99.94
CAS 7789-00-6
CAS Max % 0.06
Name Note For Silica
Linear Formula K2CrO4
Formula Weight 194.2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC188401

 
lab chem
LC188401
500mL Each for $46.37
Description & Specifications

Specifications

Formula Weight 194.2
Storage Information Room Temperature
Linear Formula K2CrO4
Specific Gravity 1g/mL
Solubility Soluble in water
Physical Form Liquid
Identification Passes Test
Name Note For Silica
CAS 7732-18-5
Chemical Name or Material Potassium Chromate
Decomposition Information Contains hexavalent Chromium
Assay 0.063%
CAS Max % 99.94
Quantity 500mL
Density 1g/mL
Packaging Poly Bottle
Color Clear, yellow
Health Hazard 2 GHS H Statement
Harmful to aquatic life with long lasting effects.
CAS 7789-00-6
Health Hazard 3 GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 0.06
Safety and Handling

GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful to aquatic life with long lasting effects.