Please login to your online account to display your discounted pricing

Potassium Chromate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$263.20 - $1,152.63

Chemical Identifiers

Chemical Name or Material Potassium Chromate
CAS 7789-00-6
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula K2CrO4
Formula Weight 194.19
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003747

 
avantor performance
305805
Poly Bottle 2.5kg Each for $1,152.63

02003748

 
avantor performance
305801
Poly Bottle 500g Each for $263.20
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 194.19
Specific Gravity 1L = 2.73kg
Chemical Name or Material Potassium Chromate
Packaging Poly Bottle
Molecular Formula K2CrO4
CAS 7789-00-6
Quantity 2.5kg