Please login to your online account to display your discounted pricing

Potassium Chromate, ACS Grade, ≥ 99.0%, LabChem™

$67.26 - $67.26

Chemical Identifiers

Chemical Name or Material Potassium Chromate
CAS 7789-00-6
CAS Max % 100
Purity Grade ACS Grade
Linear Formula K2CrO4
Formula Weight 194.2
UN Number UN3087
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC188309

 
lab chem
LC188309
100g Each for $67.26
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 194.2
Quantity 100g
UN Number UN3087
CAS 7789-00-6
Purity Grade ACS Grade
CAS Max % 100
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Collect spillage.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Chemical Name or Material Potassium Chromate
Storage Information Room Temperature
Color Yellow
Melting Point 975°C
EINECS Number 232-140-5
Solubility Soluble in water
Linear Formula K2CrO4
Physical Form Powder
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Contains hexavalent Chromium
Health Hazard 2 GHS H Statement
Causes skin irritation and serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
May cause genetic defects.
May cause cancer.
May cause damage to organs (blood, liver, kidneys) through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Collect spillage.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation and serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
May cause genetic defects.
May cause cancer.
May cause damage to organs (blood, liver, kidneys) through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.