missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Chromate, 5% (w/v), Chloride Free, Indicator for Argentometric Titrations, Ricca Chemical
$71.25 - $693.91
Chemical Identifiers
CAS | 7789-00-6 |
---|---|
Molecular Formula | CrK2O4 |
Molecular Weight (g/mol) | 194.19 |
MDL Number | MFCD00011368 |
InChI Key | XMXNVYPJWBTAHN-UHFFFAOYSA-N |
Synonym | potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687 |
PubChem CID | 24597 |
ChEBI | CHEBI:75249 |
IUPAC Name | dipotassium dioxochromiumbis(olate) |
SMILES | [K+].[K+].[O-][Cr]([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
6000-4
|
Ricca Chemical Company
60004 |
120 mL | Natural Poly Bottle |
Each for $71.25
|
|
||||
6000-16
|
Ricca Chemical Company
600016 |
500 mL | Natural Poly Bottle |
Each for $134.34
|
|
||||
6000-32
|
Ricca Chemical Company
RI600032 |
1 L | Natural Poly Bottle |
Each for $238.66
|
|
||||
6000-1
|
Ricca Chemical Company
RI60001 |
4 L | Natural Poly Bottle |
Each for $693.91
|
|
||||
Chemical Identifiers
7789-00-6 | |
194.19 | |
XMXNVYPJWBTAHN-UHFFFAOYSA-N | |
24597 | |
dipotassium dioxochromiumbis(olate) |
CrK2O4 | |
MFCD00011368 | |
potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687 | |
CHEBI:75249 | |
[K+].[K+].[O-][Cr]([O-])(=O)=O |
Specifications
7789-00-6,7732-18-5,7761-88-8 | |
4.86,97.0,7.0E-4 | |
MFCD00011368 | |
XMXNVYPJWBTAHN-UHFFFAOYSA-N | |
dipotassium dioxochromiumbis(olate) | |
24597 | |
5% (w/v) Aqueous Solution | |
120 mL |
4.67,93.0,7.0E-4 | |
CrK2O4 | |
potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687 | |
[K+].[K+].[O-][Cr]([O-])(=O)=O | |
194.19 | |
CHEBI:75249 | |
Natural Poly Bottle | |
Potassium Chromate |