Please login to your online account to display your discounted pricing

Potassium Chromate, 0.063% (w/v) Aqueous Solution, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Potassium Chromate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5990-16

 
ricca chemical company
RI599016
Plastic 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

5990-32

 
ricca chemical company
RI599032
Plastic 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

5990-4

 
ricca chemical company
RI59904
Plastic 120mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 500mL
Chemical Name or Material Potassium Chromate
Methods APHA/EPA for Silica
Packaging Plastic
Concentration or Composition (by Analyte or Components) 0.063% (w/v) Aqueous Solution