Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium chlorochromate, 97+%

Chemical Identifiers

Chemical Name or Material Potassium chlorochromate
CAS 16037-50-6
Molecular Formula ClCrKO3
Linear Formula KClCrO3
Formula Weight 174.55
MDL Number MFCD00064578
UN Number UN1479
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1900022

 
alfa aesar™
A1900022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 174.55
Odor Odorless
Assay ≥97%
Molecular Formula ClCrKO3
Linear Formula KClCrO3
CAS 16037-50-6
UN Number UN1479
Quantity 100g
Health Hazard 3 GHS P Statement
P221-P273-P201-P309-P310-P501a
Take any precaution to avoid mixing with combustibles.
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
Chemical Name or Material Potassium chlorochromate
TSCA TSCA
MDL Number MFCD00064578
Melting Point 290° to 292°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H272-H350-H317
May intensify fire; oxidizer.
May cause cancer.
May cause an allergic skin reaction.
Safety and Handling

Danger

GHS P Statement
P221-P273-P201-P309-P310-P501a
Take any precaution to avoid mixing with combustibles.
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H350-H317
May intensify fire; oxidizer.
May cause cancer.
May cause an allergic skin reaction.