Please login to your online account to display your discounted pricing

Potassium chloride, ultra dry, 99.998% (metals basis), Alfa Aesar™

$136.97 - $565.04

Chemical Identifiers

Chemical Name or Material Potassium chloride
CAS 7447-40-7
Physical Form Beads
Assay Percent Range 99.998% (metals basis)
Molecular Formula KCl
Formula Weight 74.55
MDL Number MFCD00011360
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1368209

  SDS
alfa aesar
1368209
10g Each for $136.97

AA1368218

  SDS
alfa aesar
1368218
50g Each for $565.04
Description & Specifications

Specifications

Physical Form Beads
Formula Weight 74.55
Quantity 10g
Color White
Density 1.984g/cm3
Chemical Name or Material Potassium chloride
EINECS Number 231-211-8
Odor Odorless
Assay Percent Range 99.998% (metals basis)
Molecular Formula KCl
Appearance White beads
MDL Number MFCD00011360
CAS 7447-40-7
Health Hazard 1 H303: May be harmful if swallowed.
Safety and Handling

H303: May be harmful if swallowed.