missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium chloride, ultra dry, 99.95% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1446609

 View more versions of this product

Catalog No. AA1446609



Specifications

Specifications

Potassium chloride
99.95% (metals basis)
MFCD00011360
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
[Cl-].[K+]
74.548
CHEBI:32588
Beads
White
7447-40-7
ClK
White beads
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium;chloride
4873
74.55
Odorless
10g

Chemical Identifiers

7447-40-7
74.548
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
potassium;chloride
ClK
MFCD00011360
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
CHEBI:32588
[Cl-].[K+]
Safety and Handling

Safety and Handling

H303: May be harmful if swallowed.

Documents