Please login to your online account to display your discounted pricing

Potassium chloride, ultra dry, 99.95% (metals basis), Alfa Aesar™

$81.22 - $1,093.91

Chemical Identifiers

Chemical Name or Material Potassium chloride
CAS 7447-40-7
Physical Form Beads
Assay Percent Range 99.95% (metals basis)
Molecular Formula KCl
Formula Weight 74.55
MDL Number MFCD00011360
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1446609

  SDS
alfa aesar
1446609
10g Each for $81.22

AA1446618

  SDS
alfa aesar
1446618
50g Each for $268.20

AA1446630

  SDS
alfa aesar
1446630
250g Each for $1,093.91
Description & Specifications

Specifications

Physical Form Beads
Formula Weight 74.55
Quantity 10g
Color White
Molecular Formula KCl
Assay Percent Range 99.95% (metals basis)
Appearance White beads
CAS 7447-40-7
Health Hazard 1 H303: May be harmful if swallowed.
MDL Number MFCD00011360
Chemical Name or Material Potassium chloride
Odor Odorless
Safety and Handling

H303: May be harmful if swallowed.