missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Chloride Reference Solution B, 12,856 μS/cm at 25°C, 0.1 M, Ricca Chemical
$167.50 - $276.94
Specifications
Chemical Name or Material | Potassium Chloride Reference Solution B |
---|---|
Packaging | Natural Poly Bottle |
Concentration or Composition (by Analyte or Components) | 0.1M |
Methods | ASTM D 1125 for Conductivity |
Specifications
Potassium Chloride Reference Solution B | |
Natural Poly Bottle | |
0.1M | |
ASTM D 1125 for Conductivity |