Please login to your online account to display your discounted pricing

Potassium Chloride Reference Solution A, Ricca Chemical

$95.12 - $95.12

Chemical Identifiers

Chemical Name or Material Potassium Chloride Reference Solution A
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5893-16

 
ricca chemical company
RI589316
Plastic 500mL Each for $95.12

5893-32

 
ricca chemical company
RI589332
Plastic 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 500mL
Methods ASTM D 1125 for Conductivity
Chemical Name or Material Potassium Chloride Reference Solution A
Concentration or Composition (by Analyte or Components) 1M
Packaging Plastic