missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Chloride, Grade, 99.5% min, MilliporeSigma™
Supplier: Technidata 1049365000
Specifications
Potassium Chloride | |
White | |
Crystalline Solid | |
99.5% min. | |
KCl | |
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
[Cl-].[K+] | |
74.55 | |
CHEBI:32588 | |
1.98g/cm3 (20°C); Bulk Density: 1000 kg/m3 |
EMSURE™, For Analysis | |
7447-40-7 | |
5 kg | |
ClK | |
MFCD00011360 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
potassium chloride | |
4873 | |
74.55 |
Chemical Identifiers
7447-40-7 | |
74.55 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
4873 | |
potassium chloride |
ClK | |
MFCD00011360 | |
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
CHEBI:32588 | |
[Cl-].[K+] |
Safety and Handling
Recommended Storage : Store without restrictions