missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium chloride, extra pure, ACROS Organics™

Manufacturer:  Acros Organics AC193780010

 View more versions of this product

Catalog No. AC193780010



Specifications

Specifications

Potassium chloride
10ppm max.
KCl
06,480
chlorvescent
[Cl-].[K+]
74.548
CHEBI:32588
Crystals or Crystalline Powder
Extra Pure
20ppm max.
Plastic bottle
Solubility in water: 340g/L (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and acetone
1500.0°C
White
5.0 to 8.5 (5%, H2O, 25°C)
7447-40-7
ClK
MFCD00011360
15, 7742
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium;chloride
4873
74.54
≥99.0%
50ppm max.
50ppm max.
0.1% max.
100ppm max.
1420.0°C
770.0°C
1kg

Chemical Identifiers

7447-40-7
74.548
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
potassium;chloride
ClK
MFCD00011360
chlorvescent
CHEBI:32588
[Cl-].[K+]
SDS Documents