missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Chloride (Crystalline/USP/FCC), Fisher Chemical™

Supplier:  Thermo Fisher Scientific P3303

 View more versions of this product

Catalog No. P330-3




Description

Description

Specifications

Specifications

1420°C
770°C
White
Solid
ClK
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
[Cl-].[K+]
74.55
CHEBI:32588
≥99.0% (FCC)
Pass Test (FCC), Pass Test (USP), Pass Test (USP/FCC)
Poly Bottle
Potassium Chloride
7447-40-7
6
3 kg
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride
4873
74.55
FCC/USP
1.0% max. (FCC), 1.0% max. (USP), 1.0% max. (USP/FCC)

Chemical Identifiers

7447-40-7
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
potassium chloride
ClK
MFCD00011360
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
CHEBI:32588
[Cl-].[K+]
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:0
Instability:1

SDS Documents

Documents

Promotions

Promotions