missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Chloride (Crystalline/USP/FCC/EP/BP/JP), Fisher Chemical

Manufacturer:  Thermo Scientific P33512

 View more versions of this product

Catalog No. P33512



Specifications

Specifications

Potassium Chloride
Pass Test (BP), Pass Test (EP), Pass Test (FCC), Pass Test (JP), Pass Test (USP), Pass Test (USP/FCC/EP/BP/JP)
200ppm max. (EP)
ClK
Pass Test (BP), Pass Test (EP), Pass Test (EP/BP)
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
[Cl-].[K+]
74.548
CHEBI:32588
Solid
≥99.0% (FCC)
Pass Test (USP)
Pass Test (FCC)
Pass Test (JP)
Report EU/mg (USP)
0.001% max. (USP), 0.001% max. (USP), 5mg/kg max. (FCC), 5mg/kg max. (FCC)
Pass Test (BP), Pass Test (EP), Pass Test (EP/BP/JP), Pass Test (JP)
0.5% max. (JP), 1.0% max. (BP), 1.0% max. (EP), 1.0% max. (FCC), 1.0% max. (USP), 1.0% max. (USP/EP/BP/FCC), 0.5% max. (JP)
Meets Requirements (BP), Meets Requirements (EP), Meets Requirements (USP), Meets Requirements (USP/BP/EP)
300ppm max. (BP), 300ppm max. (EP), 300ppm max. (EP/BP)
White
6
7447-40-7
200ppm max. (BP)
1ppm max. (BP), 1ppm max. (EP), 1ppm max. (USP), 1ppm max. (USP/EP/BP)
MFCD00011360
2ppm max. (JP)
Report I.U./mg (EP)
Pass Test (BP), Pass Test (EP), Pass Test (EP/BP)
potassium;chloride
4873
74.55
0.1% max. (EP), Pass Test (JP)
BP/EP/FCC/JP/USP
0.1% max. (BP)
Pass Test (USP)
Pass Test (JP)
10ppm max. (BP), 10ppm max. (EP), 10ppm max. (EP/BP), 5ppm max. (JP), 5ppm max. (JP)
Pass Test (BP), Pass Test (EP), Pass Test (FCC), Pass Test (JP), Pass Test (USP), Pass Test (USP/FCC/EP/BP/JP)
20ppm max. (BP), 20ppm max. (EP), 20ppm max. (EP/BP)
Poly Pail
0.1% max. (BP), 0.1% max. (EP), Pass Test (FCC), Pass Test (JP), Pass Test (USP), Pass Test (USP/FCC/JP), 0.1% max. (EP/BP)
1420°C
770°C
12kg

Chemical Identifiers

7447-40-7
74.548
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
potassium;chloride
ClK
MFCD00011360
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
CHEBI:32588
[Cl-].[K+]
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:0
Instability:1

SDS Documents