missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Chloride (Crystalline/Certified ACS), Fisher Chemical

Potassium chloride for calomel cells

Manufacturer:  Thermo Scientific FLP217250LB

 View more versions of this product

Catalog No. P217-250LB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Potassium Chloride
99.0 to 100.5 %
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
[Cl-].[K+]
74.548
CHEBI:32588
Solid
99.0 to 100.5%
0.002% max.
5ppm max.
0.005% max.
2ppm max.
Fiber Drum
0.005% max.
1420°C
770°C
250 lb.
7447-40-7
ClK
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
P.T. (about 0.001%)
potassium;chloride
4873
74.55
0.01% max.
Certified ACS
0.003% max.
Pass Test
0.002% max.
0.001% max.
5ppm max.
0.001% max.
White
6

Chemical Identifiers

7447-40-7
74.548
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
potassium;chloride
ClK
MFCD00011360
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
CHEBI:32588
[Cl-].[K+]
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:0
Instability:1

SDS Documents