Please login to your online account to display your discounted pricing

Potassium Chloride, Crystal, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

$191.50 - $191.50

Chemical Identifiers

Chemical Name or Material Potassium Chloride
CAS 7447-40-7
Purity Grade Ultrapure Bioreagent
Molecular Formula KCl
Formula Weight 74.55g/mol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003744

 
avantor performance
4001-06
Poly Pail 5kg Each for $191.50
Description & Specifications

Specifications

Packaging Poly Pail
Specific Gravity 1L = 1.99kg
Quantity 5kg
Chemical Name or Material Potassium Chloride
CAS 7447-40-7
Purity Grade Ultrapure Bioreagent
Physical Form Crystal
Molecular Formula KCl
Formula Weight 74.55g/mol

    For Molecular Biology Applications