Please login to your online account to display your discounted pricing

Potassium Chloride, Crystal, U.S.P. - F.C.C., J.T.Baker™

$188.71 - $1,241.74

Chemical Identifiers

Chemical Name or Material Potassium Chloride
CAS 7447-40-7
Purity Grade FCC/USP
Molecular Formula KCl
Formula Weight 74.55
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003739

 
avantor performance
304605
Poly Bottle 2.5kg Each for $1,241.74

02003742

 
avantor performance
304601
Poly Bottle 500g Each for $188.71
Description & Specifications

Specifications

Purity Grade FCC/USP
Physical Form Crystals
Formula Weight 74.55
Chemical Name or Material Potassium Chloride
CAS 7447-40-7
Packaging Poly Bottle
Specific Gravity 1L = 1.99kg
Molecular Formula KCl
Quantity 2.5kg