Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium chloride crystal optic rectangle, 38.5mm x 19.5mm x 4mm, (drilled), polished both sides

Chemical Identifiers

Chemical Name or Material Potassium chloride crystal optic rectangle
Name Note 38.5mm x 19.5mm x 4mm, (drilled), polished both sides
MDL Number MFCD00011360
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38474KT

 
alfa aesar™
38474KT
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Potassium chloride crystal optic rectangle
TSCA TSCA
Name Note 38.5mm x 19.5mm x 4mm, (drilled), polished both sides
MDL Number MFCD00011360
Quantity 1 each