Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium chloride crystal optic rectangle, 30mm x 15mm x 4mm, unpolished

Spectroscopy, IR window

Chemical Identifiers

Chemical Name or Material Potassium chloride crystal optic rectangle
Name Note 30mm x 15mm x 4mm, Unpolished
MDL Number MFCD00011360
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38401KT

 
alfa aesar™
38401KT
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

MDL Number MFCD00011360
TSCA TSCA
Name Note 30mm x 15mm x 4mm, Unpolished
Chemical Name or Material Potassium chloride crystal optic rectangle
Quantity 1 each