missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium chloride crystal optic disc, 25mm x 5mm, unpolished

Chemical Identifiers

MDL Number MFCD00011360
Chemical Name or Material Potassium chloride crystal optic disc, 25mm x 5mm, Unpolished
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA38388KT Alfa Aesar™
38388KT
1 each N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

MFCD00011360
Potassium chloride crystal optic disc, 25mm x 5mm, Unpolished

Specifications

1 each
MFCD00011360
Potassium chloride crystal optic disc, 25mm x 5mm, Unpolished

Safety and Handling

TSCA : TSCA