missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Chloride Conductivity Standard, 50μS/cm at 25°C, Ricca Chemical

Manufacturer:  Ricca Chemical Company RI5886532

 View more versions of this product

Catalog No. 13-622-605



Specifications

Specifications

Potassium Chloride Conductivity Standard
7447-40-7,7732-18-5
100.0,0.0026
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium;chloride
4873
Liquid
Colorless
50μS/cm at 25°C
98.0,0.0025
ClK
[Cl-].[K+]
74.548
CHEBI:32588
Poly Natural
1L

Chemical Identifiers

7447-40-7
74.548
4873
potassium;chloride
ClK
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
CHEBI:32588
[Cl-].[K+]
Safety and Handling

Safety and Handling

ShelfLife : 12 months

Documents